Merck
  • Determination of residues of three triphenylmethane dyes and their metabolites (malachite green, leuco malachite green, crystal violet, leuco crystal violet, and brilliant green) in aquaculture products by LC/MS/MS: first action 2012.25.
Determination of residues of three triphenylmethane dyes and their metabolites (malachite green, leuco malachite green, crystal violet, leuco crystal violet, and brilliant green) in aquaculture products by LC/MS/MS: first action 2012.25.
Journal of AOAC International (2013-11-29)
Dominique Hurtaud-Pessel, Pierrick Couëdor, Eric Verdon, Dawn Dowell
ZUSAMMENFASSUNG
During the AOAC Annual Meeting held from September 30 to October 3, 2012 in Las Vegas, NV, the Expert Review Panel (ERP) on Veterinary Drug Residues reviewed data for the method for determination of residues of three triphenylmethane dyes and their metabolites (malachite green, leuco malachite green, crystal violet, leuco crystal violet, and brilliant green) in aquaculture products by LC/MS/MS, previously published in the Journal of Chromatography A 1218, 1632-1645 (2006). The method data were reviewed and compared to the standard method performance requirements (SMPRs) found in SMPR 2009.001, published in AOAC's Official Methods of Analysis, 19th Ed. (2012). The ERP determined that the data were acceptable, and the method was approved AOAC Official First Action. The method uses acetonitrile to isolate the analyte from the matrix. Then determination is conducted by LCIMS/MS with positive electrospray ionization. Accuracy ranged from 100.1 to 109.8% for samples fortified at levels of 0.5, 0.75, 1.0, and 2.0 microg/kg. Precision ranged from 2.0 to 10.3% RSD for the intraday samples and 1.9 to 10.6% for the interday samples analyzed over 3 days. The described method is designed to accurately operate in the analytical range from 0.5 to 2 microg/kg, where the minimum required performance limit for laboratories has been fixed in the European Union at 2.0 microg/kg for these banned substances and their metabolites. Upper levels of concentrations (1-100 microg/kg) can be analyzed depending on the different optional calibrations used.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kristallviolett-Lösung -Lösung, 1%, aqueous solution
Sigma-Aldrich
Kristallviolett, certified by the BSC
Sigma-Aldrich
Kristallviolett, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Malachitgrün Oxalat, certified by the BSC
Sigma-Aldrich
Kristallviolett-Lösung -Lösung
Sigma-Aldrich
Kristallviolett, suitable for microscopy
Sigma-Aldrich
Brillantgrün, Dye content ≥85 %
Sigma-Aldrich
Malachitgrün Oxalat, suitable for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Triphenylmethan, 99%
Sigma-Aldrich
Kristallviolett-Lösung -Lösung
Sigma-Aldrich
Malachitgrün Oxalat, Technical grade
Supelco
Malachitgrünchlorid, analytical standard
Sigma-Aldrich
Malachitgrün -Lösung, suitable for microscopy
Sigma-Aldrich
Brillantgrün, certified by the BSC
Sigma-Aldrich
Kristallviolett, S. No.: 785
Sigma-Aldrich
Kristallviolett, meets USP testing specifications
Sigma-Aldrich
Leucomalachitgrün, powder
Supelco
Kristallviolett, VETRANAL®, analytical standard
Supelco
Malachitgrün Oxalat, VETRANAL®, analytical standard
Sigma-Aldrich
Schaeffer und Fulton-Sporen-Färbelösung A, suitable for microscopy
Supelco
Leucomalachitgrün, analytical standard