Merck
  • Consensus statement on the diagnosis, treatment and follow-up of patients with primary adrenal insufficiency.
Consensus statement on the diagnosis, treatment and follow-up of patients with primary adrenal insufficiency.
Journal of internal medicine (2013-12-18)
E S Husebye, B Allolio, W Arlt, K Badenhoop, S Bensing, C Betterle, A Falorni, E H Gan, A-L Hulting, A Kasperlik-Zaluska, O Kämpe, K Løvås, G Meyer, S H Pearce
ZUSAMMENFASSUNG
Primary adrenal insufficiency (PAI), or Addison's disease, is a rare, potentially deadly, but treatable disease. Most cases of PAI are caused by autoimmune destruction of the adrenal cortex. Consequently, patients with PAI are at higher risk of developing other autoimmune diseases. The diagnosis of PAI is often delayed by many months, and most patients present with symptoms of acute adrenal insufficiency. Because PAI is rare, even medical specialists in this therapeutic area rarely manage more than a few patients. Currently, the procedures for diagnosis, treatment and follow-up of this rare disease vary greatly within Europe. The common autoimmune form of PAI is characterized by the presence of 21-hydroxylase autoantibodies; other causes should be sought if no autoantibodies are detected. Acute adrenal crisis is a life-threatening condition that requires immediate treatment. Standard replacement therapy consists of multiple daily doses of hydrocortisone or cortisone acetate combined with fludrocortisone. Annual follow-up by an endocrinologist is recommended with the focus on optimization of replacement therapy and detection of new autoimmune diseases. Patient education to enable self-adjustment of dosages of replacement therapy and crisis prevention is particularly important in this disease. The authors of this document have collaborated within an EU project (Euadrenal) to study the pathogenesis, describe the natural course and improve the treatment for Addison's disease. Based on a synthesis of this research, the available literature, and the views and experiences of the consortium's investigators and key experts, we now attempt to provide a European Expert Consensus Statement for diagnosis, treatment and follow-up.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison -Lösung, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Supelco
Cortisol -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Prednisolon, ≥99%
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kortison, ≥95%
Supelco
Prednisolon, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kortison-21-Acetat, ≥99%
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Prednisolon für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisolon, VETRANAL®, analytical standard
Prednisolon, European Pharmacopoeia (EP) Reference Standard
Prednisolon für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Cortisonacetat, European Pharmacopoeia (EP) Reference Standard