Merck
  • Nandrolone decanoate inhibits gluconeogenesis and decreases fasting glucose in Wistar male rats.
Nandrolone decanoate inhibits gluconeogenesis and decreases fasting glucose in Wistar male rats.
The Journal of endocrinology (2014-01-10)
Stephan Pinheiro Frankenfeld, Leonardo Pires de Oliveira, Daniele Leão Ignacio, Raquel Guimarães Coelho, Mariana Nigro Mattos, Andrea Claudia Freitas Ferreira, Denise Pires Carvalho, Rodrigo Soares Fortunato
ZUSAMMENFASSUNG
The use of anabolic-androgenic steroids to improve physical performance or appearance has increased notably. The doses used are 10- to 100- fold higher than the therapeutic dose (TD), and this abuse can cause several side effects. Glucose metabolism is significantly affected by anabolic-androgenic steroid abuse, but studies about glycemic regulation during fasting are scarce. There are some evidences showing that testosterone can antagonize glucocorticoids action, which are crucial to glucose production during fasting. Thus, the aim of this study was to determine the impact of supraphysiological doses (SDs) of nandrolone decanoate (DECA) on rat glucose metabolism during fasting. Male Wistar rats were treated with i.m. injections of vehicle, a low TD (0.016 mg/100 g b.w.-TD group) or a high SD (1 mg/100 g b.w.-SD group) of DECA, once a week for 8 weeks. After 12 h fasting, we evaluated glucose and pyruvate tolerance tests, liver glycogen content, serum levels of gluconeogenic substrates, insulin and corticosterone, glucose uptake and hexokinase (HK) activity in skeletal muscle, and the adrenal catecholamine content. SD group had increased serum insulin levels and a blunted response to insulin regarding glucose uptake in skeletal muscle. Fasting serum glucose decreased significantly in SD group, as well as the pyruvate tolerance test and liver glycogen content. Moreover, serum levels of glycerol were increased in SD group. Our data indicate that SDs of DECA exert effects on different regulatory points of glucose metabolism, resulting in defective gluconeogenesis and decreased skeletal muscle glucose uptake in response to insulin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glukose -Lösung, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Phosphatasehemmer-Cocktail 3, DMSO solution
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose -Lösung, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
Testosteron, purum, ≥99.0% (HPLC)
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glukose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glukose, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glukose -Lösung, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Testosteron, VETRANAL®, analytical standard
Sigma-Aldrich
19-Nortestosteron, ≥99.0% (HPLC)
Testosteron, European Pharmacopoeia (EP) Reference Standard
Nandrolon-decanoat, European Pharmacopoeia (EP) Reference Standard
Nandrolon decanoat für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Nandrolondecanoat für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard