Merck
  • [Removal of heavy metals from extract of Angelica sinensis by EDTA-modified chitosan magnetic adsorbent].
[Removal of heavy metals from extract of Angelica sinensis by EDTA-modified chitosan magnetic adsorbent].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2014-02-06)
Yong Ren, Ming-Hui Sun, Hong Peng, Kai-Xun Huang
ZUSAMMENFASSUNG
The concentrations of heavy metals in the extracting solutions of traditional Chinese medicine are usually very low. Furthermore, a vast number of organic components contained in the extracting solutions would be able to coordinate with heavy metals, which might lead to great difficulty in high efficient removal of them from the extracting solutions. This paper was focused on the removal of heavy metals of low concentrations from the extracting solution of Angelica sinensis by applying an EDTA-modified chitosan magnetic adsorbent (EDTA-modified chitosan/SiO2/Fe3O4, abbreviated as EDCMS). The results showed that EDCMS exhibited high efficiency for the removal of heavy metals, such as Cu, Cd and Pb, e.g. the removal percentage of Cd and Pb reached 90% and 94.7%, respectively. Besides, some amounts of other heavy metals like Zn and Mn were also removed by EDCMS. In addition, the total solid contents, the amount of ferulic acid and the HPLC fingerprints of the extracting solution were not changed significantly during the heavy metal removal process. These results indicate that EDCMS may act as an applicable and efficient candidate for the removal of heavy metals from the extracting solution of A. sinensis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Supelco
Dinatriumedetat Dihydrat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure Calcium-Dinatriumsalz Hydrat
Sigma-Aldrich
Chitosan aus Garnelenschalen, low-viscous
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, 2%, solution
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ≥97.0% (KT)