Merck
  • Optimization of complex fermentation media for glucose oxidase production using statistical approach.
Optimization of complex fermentation media for glucose oxidase production using statistical approach.
Pakistan journal of biological sciences : PJBS (2014-02-08)
Ahmad Anas Nagoor Gunny, Dachyar Arbain, Logachanthirika Sithamparam
ZUSAMMENFASSUNG
Production cost of enzyme is largely determined by the type of the strain and raw material used to propagate the strain. Hence, selection of the strain and raw materials is crucial in enzyme production. For Glucose oxidase (GOx), previous studies showed Aspergillus terreus UniMAP AA-1 offers a better alternative to the existing sources. Thus, a lower production cost could be logically anticipated by growing the strain in a cheaper complex media such as molasses. In this work, sugar cane molasses, supplemented with urea and carbonate salt and a locally isolated strain Aspergillus terreus UniMAP AA-1 were used to produce a crude GOx enzyme in a small scale. A statistical optimization approach namely Response Surface Methodology (RSM) was used to optimize the media components for highest GOx activity. It was found that the highest GOx activity was achieved using a combination of molasses, carbonate salt and urea at concentration 32.51, 4.58 and 0.93% (w/v), respectively. This study provides an alternative optimized media conditions for GOx production using locally available raw materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Calciumcarbonat, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
D-(+)-Glukose -Lösung, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Glucose-Oxidase aus Aspergillus niger, Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Glucose-Oxidase aus Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
D-(+)-Glukose -Lösung, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Calciumcarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
D-(+)-Gluconsäure δ-Lacton, ≥99.0% (GC)
Sigma-Aldrich
Calciumcarbonat, ReagentPlus®
Sigma-Aldrich
Calciumcarbonat, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calciumcarbonat, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Supelco
Calciumcarbonat, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)