Merck
  • Mercury transportation in soil via using gypsum from flue gas desulfurization unit in coal-fired power plant.
Mercury transportation in soil via using gypsum from flue gas desulfurization unit in coal-fired power plant.
Journal of environmental sciences (China) (2014-02-14)
Kelin Wang, William Orndorff, Yan Cao, Weiping Pan
ZUSAMMENFASSUNG
The mercury flux in soils was investigated, which were amended by gypsums from flue gas desulphurization (FGD) units of coal-fired power plants. Studies have been carried out in confined greenhouses using FGD gypsum treated soils. Major research focus is uptakes of mercury by plants, and emission of mercury into the atmosphere under varying application rates of FGD gypsum, simulating rainfall irrigations, soils, and plants types. Higher FGD gypsum application rates generally led to higher mercury concentrations in the soils, the increased mercury emissions into the atmosphere, and the increased mercury contents in plants (especially in roots and leaves). Soil properties and plant species can play important roles in mercury transports. Some plants, such as tall fescue, were able to prevent mercury from atmospheric emission and infiltration in the soil. Mercury concentration in the stem of plants was found to be increased and then leveled off upon increasing FGD gypsum application. However, mercury in roots and leaves was generally increased upon increasing FGD gypsum application rates. Some mercury was likely absorbed by leaves of plants from emitted mercury in the atmosphere.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Drierit, with indicator, 8 mesh
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Drierit, with indicator, 10-20 mesh
Sigma-Aldrich
Calciumsulfat Dihydrat, ReagentPlus®, ≥99%
Sigma-Aldrich
Calciumsulfat, −325 mesh, 99%
Sigma-Aldrich
Schwefel, powder, 99.98% trace metals basis
Sigma-Aldrich
Schwefel, 99.998% trace metals basis
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Calciumsulfat Dihydrat, ACS reagent, 98%
Sigma-Aldrich
Drierit, with indicator, 4 mesh
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Quecksilber, ACS reagent, 99.9995% trace metals basis
Sigma-Aldrich
Wasser, ACS reagent
Sigma-Aldrich
Drierit, with indicator, 6 mesh
Supelco
Wasser, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Calciumsulfat Dihydrat, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calciumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Schwefel, flakes, ≥99.99% trace metals basis
Supelco
Schwefel, PESTANAL®, analytical standard