Merck
Somatic RHOA mutation in angioimmunoblastic T cell lymphoma.
Nature genetics (2014-01-15)
Mamiko Sakata-Yanagimoto, Terukazu Enami, Kenichi Yoshida, Yuichi Shiraishi, Ryohei Ishii, Yasuyuki Miyake, Hideharu Muto, Naoko Tsuyama, Aiko Sato-Otsubo, Yusuke Okuno, Seiji Sakata, Yuhei Kamada, Rie Nakamoto-Matsubara, Nguyen Bich Tran, Koji Izutsu, Yusuke Sato, Yasunori Ohta, Junichi Furuta, Seiichi Shimizu, Takuya Komeno, Yuji Sato, Takayoshi Ito, Masayuki Noguchi, Emiko Noguchi, Masashi Sanada, Kenichi Chiba, Hiroko Tanaka, Kazumi Suzukawa, Toru Nanmoku, Yuichi Hasegawa, Osamu Nureki, Satoru Miyano, Naoya Nakamura, Kengo Takeuchi, Seishi Ogawa, Shigeru Chiba
ZUSAMMENFASSUNG
Angioimmunoblastic T cell lymphoma (AITL) is a distinct subtype of peripheral T cell lymphoma characterized by generalized lymphadenopathy and frequent autoimmune-like manifestations. Although frequent mutations in TET2, IDH2 and DNMT3A, which are common to various hematologic malignancies, have been identified in AITL, the molecular pathogenesis specific to this lymphoma subtype is unknown. Here we report somatic RHOA mutations encoding a p.Gly17Val alteration in 68% of AITL samples. Remarkably, all cases with the mutation encoding p.Gly17Val also had TET2 mutations. The RHOA mutation encoding p.Gly17Val was specifically identified in tumor cells, whereas TET2 mutations were found in both tumor cells and non-tumor hematopoietic cells. RHOA encodes a small GTPase that regulates diverse biological processes. We demonstrated that the Gly17Val RHOA mutant did not bind GTP and also inhibited wild-type RHOA function. Our findings suggest that impaired RHOA function in cooperation with preceding loss of TET2 function contributes to AITL-specific pathogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isocitronen-Dehydrogenase (NADP) aus Schweineherz, Type I, 0.5-3.0 unit/mg solid (plus numerous enzyme activities associated with porcine heart)
Sigma-Aldrich
Isocitronen-Dehydrogenase (NADP) aus Schweineherz, Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in E. coli, lyophilized powder, ≥80 units/mg protein
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in Sf9 cells, ≥90% (SDS-PAGE)