Merck
  • Direct detection of neuropeptide dynorphin A binding to the second extracellular loop of the κ opioid receptor using a soluble protein scaffold.
Direct detection of neuropeptide dynorphin A binding to the second extracellular loop of the κ opioid receptor using a soluble protein scaffold.
The FEBS journal (2014-03-13)
Johannes Björnerås, Martin Kurnik, Mikael Oliveberg, Astrid Gräslund, Lena Mäler, Jens Danielsson
ZUSAMMENFASSUNG
The molecular determinants for selectivity of ligand binding to membrane receptors are of key importance for the understanding of cellular signalling, as well as for rational therapeutic intervention. In the present study, we target the interaction between the κ opioid receptor (KOR) and its native peptide ligand dynorphin A (DynA) using solution state NMR spectroscopy, which is generally made difficult by the sheer size of membrane bound receptors. Our method is based on 'transplantation' of an extracellular loop of KOR into a 'surrogate' scaffold; in this case, a soluble β-barrel. Our results corroborate the general feasibility of the method, showing that the inserted receptor segment has negligible effects on the properties of the scaffold protein, at the same time as maintaining an ability to bind its native DynA ligand. Upon DynA binding, only small induced chemical shift changes of the KOR loop were observed, whereas chemical shift changes of DynA and NMR paramagnetic relaxation data show conclusively that the peptide interacts with the inserted loop. The binding interface is composed of a disordered part of the KOR loop and involves both electrostatic and hydrophobic interactions. Even so, simultaneous effects along the DynA sequence upon binding show that control of the recognition is a concerted event.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Sigma-Aldrich
Superoxid-Dismutase Rind, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxid-Dismutase aus Rinderleber, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxid-Dismutase aus Humanerythrocyten, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxid-Dismutase aus Rinderleber, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)