Sigma-Aldrich
Schwefel, powder, 99.98% trace metals basis
Sigma-Aldrich
Schwefel, 99.998% trace metals basis
Sigma-Aldrich
Silizium, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Schwefel, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Supelco
Schwefel, PESTANAL®, analytical standard
Sigma-Aldrich
Silizium, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silizium, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Silikon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Silikon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Silizium, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Silikon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Silikon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Silizium, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Silikon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Silikon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Silikon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Silikon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Silikon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Silikon, rod, 100mm, diameter 25mm, crystalline, 100%