Merck
  • High-dose busulfan-thiotepa with autologous stem cell transplantation followed by posterior fossa irradiation in young children with classical or incompletely resected medulloblastoma.
High-dose busulfan-thiotepa with autologous stem cell transplantation followed by posterior fossa irradiation in young children with classical or incompletely resected medulloblastoma.
Pediatric blood & cancer (2014-01-29)
Guillaume Bergthold, Maria El Kababri, Pascale Varlet, Frederic Dhermain, Christian Sainte-Rose, Marie-Anne Raquin, Virginie Kieffer, Gisele Goma, Jacques Grill, Dominique Valteau-Couanet, Christelle Dufour
ZUSAMMENFASSUNG
The aim of the study is to evaluate the outcome of young children with high risk localized medulloblastomas (newly diagnosed classical or incompletely resected) treated by high-dose busulfan-thiotepa with autologous stem cell rescue (ASCT) followed by focal radiation therapy (RT). Between September 1994 and January 2010, 19 children younger than 5 years old at diagnosis fulfilling the above inclusion criteria were treated at the Institute Gustave Roussy. After conventional chemotherapy, they received busulfan at a dose of 600 mg/m(2) and thiotepa at a dose of 900 mg/m(2) followed by ASCT. Focal RT was delivered at least 70 days after ASCT. The median follow-up was 40.5 months (range, 14.5-191.2 months). The 3-year event-free survival (EFS) and OS were 68% (95% CI 45-84%) and 84% (95% CI 61-94%), respectively. Acute toxicity consisted mainly in hepatic veno-occlusive disease (6/19 patients) and bone marrow aplasia (all patients). No toxic death occurred. The Full Scale Intellectual Quotient tended to decrease over time at a mean rate of 0.9 point per year from the date of diagnosis. This intensive treatment resulted in a high overall survival rate in young children with newly diagnosed non-metastatic classic or incompletely resected MB. In spite of a high incidence of hepatic veno-occlusive disease (32%), the acute toxicity was manageable. Delayed neuropsychological side effects remain main concerns. These results should to be confirmed in a larger cohort.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
1,4-Butandiol-dimethansulfonat, analytical standard, for drug analysis
Busulfan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thio-TEPA