Merck
  • Characterization of the motor inhibitory role of colonic mucosa under chemical stimulation in mice.
Characterization of the motor inhibitory role of colonic mucosa under chemical stimulation in mice.
American journal of physiology. Gastrointestinal and liver physiology (2014-02-15)
Francisco E Martín-Cano, Pedro J Camello, María J Pozo
ZUSAMMENFASSUNG
The main roles of the colonic mucosa are the absorption of water and electrolytes and the barrier function that preserves the integrity of the colonic wall. The mediators and mechanisms to accomplish these functions are under continuous investigation, but little attention has been paid to a possible control of colonic motility by the mucosa that would fine tune the relationship between absorption and motility. The purpose of this study was to establish the role of the mucosa in the control of induced colonic contractility. Young ICR-CD1 mice (3-5 mo old) were studied. Isometric tension transducers were used to record contractility in full-thickness (FT) and mucosa-free (MF) strips from proximal colon. Proximal FT strips showed lower KCl- and bethanechol-induced responses than MF strips. The difference was not due to mechanical artefacts since the contractile response of FT strips to electrical field stimulation was around 50% lower than in MF. The inhibitory effects of the mucosa on FT strips were mimicked by immersion of separate strips of mucosa in the organ bath but not by addition of mucosal extract, suggesting gaseous molecules as mediators of this effect. Incubation of MF strips with synthase inhibitors of nitric oxide, carbon monoxide, and hydrogen sulfide abolished the inhibition caused by addition of the mucosal strip, indicating that mucosal gasotransmitters are the mediators of these effects. This suggests that the control of colonic motility exerted by the mucosa could fine tune the balance between transit and absorption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Kaliumchlorid, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Kaliumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Kaliumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Potassium chloride -Lösung, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Kaliumchlorid, BioXtra, ≥99.0%
Supelco
Kaliumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kohlenstoffmonoxid, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Kaliumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Kaliumchlorid -Lösung, BioUltra, ~3 M in H2O
Sigma-Aldrich
Kaliumchlorid -Lösung, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Kaliumchlorid, ≥99.99% trace metals basis
Supelco
Kaliumchlorid -Lösung, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
ISA (Ionenstärke-Einstellreagens: 1 M KCl), 1 M KCl
Supelco
Kaliumchlorid -Lösung, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Kaliumchlorid -Lösung, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Kaliumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Kaliumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Kaliumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Carbamyl-β-Methylcholinchlorid, ≥99% (TLC), crystalline