Merck
The effect of methotrexate injection for treatment of an ectopic pregnancy on ovarian reserve.
Fertility and sterility (2014-02-19)
ChengWei Xiao, Michael H Dahan
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Amethopterin Hydrat, ≥98% (HPLC), powder
Sigma-Aldrich
L-Amethopterin Hydrat, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexat, meets USP testing specifications
SAFC
Methotrexat
Supelco
Methotrexat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methotrexat -Lösung, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Amethopterin Hydrat, ≥99.0% (sum of enantiomers, HPLC)
Methotrexat für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Methotrexat für die Systemeignung, European Pharmacopoeia (EP) Reference Standard