Merck
Synergistic cytotoxic activity of treosulfan and gemcitabine in pancreatic cancer cell lines.
Anticancer research (2014-04-03)
Emilia Nitsch, Sormeh Mina, Ingo Brammer, Andrea Pace, Gunter Schuch, Carsten Bokemeyer, Axel Zander, Nicolaus Kröger, Francis Ayuk
ZUSAMMENFASSUNG
Treatment for advanced pancreatic cancer is still very unsatisfactory. Treosulfan is an alkylating agent used for conventional, as well as high-dose chemotherapy regimens, whereby plasma concentrations over 500 μg/ml can be achieved. We investigated the effects of treosulfan on pancreatic cancer cell lines. Using Panc-1, MIA PaCa-2 and Capan-2 cell lines, we investigated the in vitro cytotoxicity of treosulfan-alone and in combination with gemcitabine, 5-fluorouracil or irradiation. Treosulfan was potently cytotoxic against all pancreatic cancer cell lines at all concentrations (1-100 μg/ml). Combination of treosulfan and gemcitabine revealed strong synergistic effects independent of the sequence of drug administration. Similarly, synergism was observed with irradiation. Combination of treosulfan and 5-fluorouracil revealed antagonism. Treosulfan effectively kills pancreatic carcinoma cells in vitro and has synergistic activity in combination with gemcitabine and irradiation. These results warrant further investigation of treosulfan in the treatment of pancreatic cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2′-Desoxycytidin, ≥99% (HPLC)
Supelco
1,4-Butandiol-dimethansulfonat, analytical standard, for drug analysis
Busulfan, European Pharmacopoeia (EP) Reference Standard