Merck
  • Whole-head washing, prior to cutting, provides sanitization advantages for fresh-cut Iceberg lettuce (Latuca sativa L.).
Whole-head washing, prior to cutting, provides sanitization advantages for fresh-cut Iceberg lettuce (Latuca sativa L.).
International journal of food microbiology (2014-04-08)
Sindy Palma-Salgado, Arne J Pearlstein, Yaguang Luo, Hee Kyung Park, Hao Feng
ZUSAMMENFASSUNG
The efficacy of two leafy produce wash methods, the traditional cutting-before-washing process and a new washing-before-cutting method, on reduction of Escherichia coli O157:H7 inoculated on Iceberg lettuce was compared. The washing tests were conducted in a pilot-scale washer using combinations of water, chlorine, peroxyacetic acid, and ultrasound. The washing-before-cutting process recorded an E. coli O157:H7 count reduction 0.79-0.80 log₁₀ CFU/g higher than that achieved with the cutting-before-washing process in treatments involving only a sanitizer. When ultrasound was applied to the washing-before-cutting process, a further improvement of 0.37-0.68 log₁₀ CFU/g in microbial count reduction was obtained, reaching total reductions of 2.43 and 2.24 log₁₀ CFU/g for chlorine and peroxyacetic acid washes, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chlor, ≥99.5%
Sigma-Aldrich
Chlor, puriss., ≥99.8%