Merck
[Cystinosis: from cystine crystals to the cystinosin].
Nefrologia : publicacion oficial de la Sociedad Espanola Nefrologia (2003-04-24)
G Pintos
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Cystin, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Cystin
Sigma-Aldrich
L-Cystin, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystin, ≥99.7% (TLC)
Supelco
L-Cystin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cystin, Wacker Chemie AG, ≥98.5%
Supelco
L-Cystin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Cystin, European Pharmacopoeia (EP) Reference Standard