Merck
  • Green tea extract thermogenesis-induced weight loss by epigallocatechin gallate inhibition of catechol-O-methyltransferase.
Green tea extract thermogenesis-induced weight loss by epigallocatechin gallate inhibition of catechol-O-methyltransferase.
Journal of medicinal food (2007-01-05)
Q Shixian, B VanCrey, J Shi, Y Kakuda, Y Jiang
ZUSAMMENFASSUNG
Epidemiological studies have shown that intake of tea catechins is associated with a lower risk of cardiovascular disease. The antioxidative activity of tea-derived catechins has been extensively studied. Reports have shown that green tea extract intake is associated with increased weight loss due to diet-induced thermogenesis, which is generally attributed to the catechin epigallocatechin gallate. That catechin-polyphenols are known to be capable of inhibiting catechol-O-methyltransferase (the enzyme that degrades norepinephrine) is a possible explanation for why the green tea extract is effective in stimulating thermogenesis by epigallocatechin gallate to augment and prolong sympathetic stimulation of thermogenesis. Knowledge about thermogenesis-induced weight loss produced by green tea's epigallocatechin gallate and its ability to inhibit catechol-O-methyltransferase is important for health benefits and for prolonging the action of norepinephrine in the synaptic cleft.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Epigallokatechingallat, ≥95%
Sigma-Aldrich
(−)-Epigallokatechingallat, ≥80% (HPLC), from green tea
Supelco
(−)-Epigallokatechingallat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallokatechingallat, analytical standard
Epigallocatechin-gallat, primary reference standard