Merck
[Admissible concentration in biological material: an index of exposure to dichloromethane].
Medycyna pracy (2008-07-26)
Małgorzata Kupczewska-Dobecka, Renata Soćko
ZUSAMMENFASSUNG
In 2007, a new maximum admissible concentration value of 88 mg/m3 was specified for dichloromethane (DCM) in the Polish list of admissible concentrations of harmful chemicals and dusts in the work environment atmosphere. The new value is four times higher than the former one (20 mg/m3), valid in Poland for 20 past years. At the same time, it was decided that the value of short term exposure limit (STEL) shall no longer be specified. Dichloromethane is a chemical which can be absorbed by routes other than respiratory, therefore, biological monitoring is required to ensure that the risk is not underestimated. Experimental data support the conclusion that volatile chloroorganic compounds, including dichloromethane, are excreted with urine in the unchanged form, and thus their absorption and exposure can be easily assessed. Concentration of dichloromethane in urine determined at the end of work shift is proposed in Poland as a specific index of dichloromethane exposure. Dichloromethane inhalation exposure at maximum admissible concentration of 88 mg/m3 corresponds with biological exposure index (BEI) of 0.15 mg DCM/l urine. The BEI value proposed in Poland for DCM is compatible with that specified by the American Conference of Governmental Industrial Hygienists (ACGIH).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichlormethan, analytical standard
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichlormethan, Selectophore, ≥99.5%