Merck
Hydroxyurea and hydroxamic acid derivatives as antitumor drugs.
Cancer chemotherapy and pharmacology (2009-04-08)
Nina Saban, Maro Bujak
ZUSAMMENFASSUNG
Hydroxyurea has been used for decades and it is still valuable for the treatment of some types of cancer. It inhibits ribonucleotide reductase (RNR) enzyme known to be crucial in the conversion of ribonucleotides into deoxyribonucleotides. However, nowadays the main focus has shifted to structurally similar hydroxamic acid derivatives that target specific enzymes involved in cancer progression such as histone deacetylases, matrix metalloproteinases and also RNR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydroxyharnstoff, 98%, powder
Hydroxycarbamid, European Pharmacopoeia (EP) Reference Standard