Merck
Ciprofloxacin: review on developments in synthetic, analytical, and medicinal aspects.
Journal of enzyme inhibition and medicinal chemistry (2010-03-20)
Prabodh Chander Sharma, Ankit Jain, Sandeep Jain, Rakesh Pahwa, Mohammad Shahar Yar
ZUSAMMENFASSUNG
In the current practices of anti-infective therapy, ciprofloxacin is a very popular fluoroquinolone having a broad spectrum of activity and diverse therapeutic prospects. The reasons for its wide use include multiresistant pathogens susceptible only to ciprofloxacin. The available clinical evidence suggests the potentially enhanced efficacy of this drug in the treatment of various community acquired and nosocomial infections, e.g. respiratory tract, urinary tract, and skin infections and sexually transmitted diseases. As compared to other agents of its class, the pharmacokinetic profile of ciprofloxacin demonstrates equivalent or greater bioavailability, higher plasma concentrations, and increased tissue penetration, as reflected in the greater volume of distribution. Various molecular modifications of this drug have been made to further improve its characteristics. Several methods of analytical determination of ciprofloxacin and its metabolites in biological fluids employing various techniques have been reported. The present article is focused on the synthetic development, pharmacotherapeutic, and analytical evaluation vistas of ciprofloxacin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard