Merck
Role of endothelin in thick ascending limb sodium chloride transport.
Contributions to nephrology (2011-09-07)
Vanesa D Ramseyer, Pablo D Cabral, Jeffrey L Garvin
ZUSAMMENFASSUNG
The thick ascending limb of the loop of Henle reabsorbs 20-30% of filtered sodium chloride (NaCl) and generates the osmotic gradient necessary for water absorption in the distal nephron. It is second only to the collecting duct as a source of renal endothelin (ET)-1, which inhibits NaCl reabsorption in the thick ascending limb by reducing NaCl entry into the cell via the furosemide-sensitive Na(+)/K(+)/2 Cl(-) cotransporter. The mechanism by which this occurs appears to be due to activation of ET(B) receptors, phosphatidylinositol 3 kinase and Akt, and enhanced nitric oxide production by nitric oxide synthase 3. ET-1 may inhibit thick ascending limb NaCl absorption in either an autocrine or paracrine fashion. High-salt intake elevates ET-1 release by thick ascending limbs, although the molecular mechanism involved is unknown. Enhanced ET-1 release and inhibition of thick ascending limb NaCl absorption are thought to be among the mechanisms required to eliminate a salt load without increasing blood pressure. However, we still have much to learn about how ET-1 inhibits thick ascending limb NaCl absorption, how release and processing of ET-1 are regulated, and the receptors involved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis