Merck
  • A nexus of progression of chronic kidney disease: tryptophan, profibrotic cytokines, and charcoal.
A nexus of progression of chronic kidney disease: tryptophan, profibrotic cytokines, and charcoal.
Journal of renal nutrition : the official journal of the Council on Renal Nutrition of the National Kidney Foundation (2011-12-28)
Gerald Schulman
ZUSAMMENFASSUNG
Fibrosis plays a major role in the pathogenesis of progressive chronic kidney disease (CKD). The inhibition of the renin-angiotensin system, which promotes fibrosis, has become the standard of care in the treatment of patients with CKD. The use of alternative agents capable of blocking the actions of profibrotic cytokines such as transforming growth factor-beta (TGF-β) is also an important strategy that is in its early stages of development. An example of such a drug is AST-120, a charcoal compound that ultimately inhibits the synthesis of TGF-β in the kidney. The inhibition is mediated by blocking the intestinal absorption of tryptophan-derived indole by AST-120. This reduces the hepatic conversion of indole to indoxyl sulfate (IS). IS stimulates the production of TGF-β in the renal parenchyma, and lowering the level of IS with AST-120 appears to slow progression of CKD. The status of recent trials examining the safety and efficacy of AST-120 has been described, including a multicenter, randomized, placebo-controlled, phase III trial of approximately 2,000 subjects being conducted to gain approval of this drug by the U.S. Food and Drug Administration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Aktivkohle, powder
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Supelco
Aktivkohle, puriss. p.a., powder
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Millipore
Aktivkohle, suitable for GC
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Kohlenstoff, foil, 10x10mm, thickness 2.0mm, hOpg
Kohlenstoff, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Kohlenstoff, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Kohlenstoff, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Kohlenstoff, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon