Merck
  • Gustatory detection of tetrodotoxin and saxitoxin, and its competitive inhibition by quinine and strychnine in freshwater fishes.
Gustatory detection of tetrodotoxin and saxitoxin, and its competitive inhibition by quinine and strychnine in freshwater fishes.
Marine drugs (2011-12-14)
Toshiaki J Hara
ZUSAMMENFASSUNG
Fish detect extremely low levels of marine toxins tetrodotoxin (TTX) and saxitoxin (STX) via the specialized gustatory receptor(s). Physiological and pharmacological studies show that receptor(s) for TTX and STX are distinct from those which detect feeding stimulant amino acids and bile acids, and that TTX and STX do not share the same receptor populations, while interacting with quinine and strychnine in a competitive fashion suggestive of an antidotal relationship.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Strychnin
Sigma-Aldrich
Chinin, 90%
Sigma-Aldrich
Chinin, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Supelco
Chinin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland