Merck
  • Towards advanced aqueous dye removal processes: a short review on the versatile role of activated carbon.
Towards advanced aqueous dye removal processes: a short review on the versatile role of activated carbon.
Journal of environmental management (2012-03-31)
Gergo Mezohegyi, Frank P van der Zee, Josep Font, Agustí Fortuny, Azael Fabregat
ZUSAMMENFASSUNG
During the last decade, several physico-chemical and biological techniques have been developed to remove colour from textile wastewaters. Some of these techniques rely on and many will profit from activated carbon (AC). The role of AC is versatile: (1) it acts as a dye adsorbent, not only in straightforward adsorption processes but also in AC-enhanced coagulation and membrane filtration processes; (2) it generates strong oxidising agents (mostly, hydroxyl (OH) radicals) in electrochemical dye oxidation; (3) it catalyses OH production in advanced oxidation processes; (4) it catalyses anaerobic (azo) dye reduction and supports biofilm growth in microbial dye removal. This paper reviews the role of AC in dye decolourisation, evaluates the feasibility of each AC-amended decolourisation technique and discusses perspectives on future research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Aktivkohle, powder
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Supelco
Aktivkohle, puriss. p.a., powder
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Millipore
Aktivkohle, suitable for GC
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Kohlenstoff, foil, 10x10mm, thickness 2.0mm, hOpg
Kohlenstoff, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Kohlenstoff, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Kohlenstoff, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Kohlenstoff, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon