- [The world of galactose and glucose: pathogenesis and therapy of lysosomal diseases].
[The world of galactose and glucose: pathogenesis and therapy of lysosomal diseases].
No to hattatsu = Brain and development (2014-04-18)
Yoshiyuki Suzuki
PMID24738180
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glukose -Lösung, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose -Lösung, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glukose -Lösung, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard