Merck
[Targeting chemotherapy with activated carbon particles adsorbing anti-cancer drugs].
Gan to kagaku ryoho. Cancer & chemotherapy (1988-04-01)
A Hagiwara, T Takahashi, S Kondoh, A Iwamoto, K Sawai
ZUSAMMENFASSUNG
A new drug delivery system comprising activated carbon particles adsorbing anticancer drugs was developed. Activated carbon particles have affinity to the regional lymph-nodes and the tumor surface. When activated carbon particles adsorbing anticancer drug were administered into the tissues or serosal cavity, they remained for a long time at the site of application, and released the drug slowly. Activated carbon particles adsorbing mitomycin C (MMC-CH) were prepared for the use of an intracavitary administration. In animal experiment tissues, MMC-CH distributed a large amount of mitomycin C to the intraperitoneal tissues, while there was a low level of mitomycin C in the whole body. The LD50 value of MMC-CH was 5.3 times as high as that of mitomycin C aqueous solution. The therapeutic index (LD50/ED50) of the MMC-CH therapy was 3.1 times higher than that of the mitomycin C aqueous solution in Yoshida sarcoma ascites. Clinical trials of MMC-CH were performed in 81 patients with malignant effusion. Fifty-one of the 81 patients responded well. Activated carbon particles adsorbing peplomycin (PEP-CH) were prepared for chemotherapy against lymphatic metastasis. A high level of peplomycin was distributed to regional lymph nodes for a long time, when it was injected into the gastric wall of dogs. In animal experiments using MH 134 tumor, the therapeutic efficacy of PEP-CH on the lymphatic metastasis was greater than that of peplomycin aqueous solution. Clinical trials were carried out in 7 patients with esophageal cancer who did not undergo surgery. The local response rate was 85% and the survival time was 9.4 months in average including 5 survivors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Aktivkohle, powder
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Supelco
Aktivkohle, puriss. p.a., powder
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Millipore
Aktivkohle, suitable for GC
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Kohlenstoff, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Kohlenstoff, foil, 10x10mm, thickness 2.0mm, hOpg
Kohlenstoff, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Kohlenstoff, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Kohlenstoff, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon