Merck
  • Agar capillary clonogenic microassays for cellular immunocytotoxic activities in human leukaemia and lymphoma.
Agar capillary clonogenic microassays for cellular immunocytotoxic activities in human leukaemia and lymphoma.
Leukemia & lymphoma (1993-03-01)
H R Maurer, H T Hassan
ZUSAMMENFASSUNG
Current concepts of immunotherapeutic approaches in leukemias and lymphomas using activated cytotoxic lymphocytes and macrophages are briefly reviewed. Defective cellular immunocytotoxic activities and effects of interleukins and chemotherapeutic drugs thereupon are discussed. In vitro assays to measure lymphokine-activated killer (LAK) and natural killer (NK) cell activities suffer from various problems, depending on the quality of the endpoints. Our clonogenic microassay for LAK cell activity, using agar-containing glass capillaries, avoids some of the potential artifacts and offers several advantages that are discussed. As an example the stimulatory effect of low mafosfamide concentrations on the LAK cell activity versus K562 human myeloid leukemia cells is demonstrated. Thus, our clonogenic LAK microassay provides a valid tool for preclinical screening of immunomodulatory agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Millipore
Agar, powder, suitable for microbiology
Sigma-Aldrich
Noble-Agar
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Bakteriologischer Agar, for molecular biology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Select-Agar
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, suitable for plant cell culture
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology