Merck
[From classical corticotherapy to cortisone loading].
Annales de medecine interne (1994-06-01)
J Arlet
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kortison, ≥95%
Supelco
Cortisone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®