- O-Acetyltransferases for chloramphenicol and other natural products.
O-Acetyltransferases for chloramphenicol and other natural products.
Antimicrobial agents and chemotherapy (1997-01-01)
I A Murray, W V Shaw
PMID8980745
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard