Merck
  • Ascorbic acid deficiency affects genes for oxidation-reduction and lipid metabolism in livers from SMP30/GNL knockout mice.
Ascorbic acid deficiency affects genes for oxidation-reduction and lipid metabolism in livers from SMP30/GNL knockout mice.
Biochimica et biophysica acta (2014-04-08)
Keita Takahashi, Yuki Kishimoto, Tomokazu Konishi, Yasunori Fujita, Masafumi Ito, Kentaro Shimokado, Naoki Maruyama, Akihito Ishigami
ZUSAMMENFASSUNG
We sought to elucidate the effect of an ascorbic acid (AA) deficiency on gene expression, because the water soluble antioxidant AA is an important bioactive substance in vivo. We performed microarray analyses of the transcriptome in the liver from senescence marker protein-30 (SMP30)/gluconolactonase (GNL) knockout (KO) mice, which are unable to synthesize AA in vivo. Our microarray analysis revealed that the AA deficiency increased gene expression related to the oxidation-reduction process, i.e., the nuclear factor, erythroid derived 2, like 2 (Nrf2) gene, which is a reactive oxygen species-sensitive transcriptional factor. Moreover, this AA deficiency increased the expression of genes for lipid metabolism including the cytochrome P450, family 7, subfamily a, polypeptide 1 (Cyp7a1), which is a late-limiting enzyme of the primary bile acid biosynthesis pathway. Although an AA deficiency increased the Cyp7a1 protein level, bile acid levels in the liver and gallbladder decreased. Since Cyp7a1 has a heme iron at the active site, AA must function as a reductant of the iron required for the continuous activation of Cyp7a1. This experimental evidence strongly supports a role for AA in the physiologic oxidation-reduction process and lipid metabolism including bile acid biosynthesis. Although many effects of AA supplementation have been reported, no microarray analysis of AA deficiency in vivo is available. Results from using this unique model of AA deficiency, the SMP30/GNL-KO mouse, now provide new information about formerly unknown AA functions that will implement further study of AA in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
(+)-Natrium-L-ascorbat, crystalline, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(+)-Natrium-L-ascorbat, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Supelco
Natriumascorbat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Natriumascorbat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.