Merck
  • Feasibility, efficacy, and adverse effects of outpatient antibacterial prophylaxis in children with acute myeloid leukemia.
Feasibility, efficacy, and adverse effects of outpatient antibacterial prophylaxis in children with acute myeloid leukemia.
Cancer (2014-03-29)
Hiroto Inaba, Aditya H Gaur, Xueyuan Cao, Patricia M Flynn, Stanley B Pounds, Viswatej Avutu, Lindsay N Marszal, Scott C Howard, Ching-Hon Pui, Raul C Ribeiro, Randall T Hayden, Jeffrey E Rubnitz
ZUSAMMENFASSUNG
Intensive chemotherapy for pediatric acute myeloid leukemia incurs the risk of infectious complications, but the benefits of antibiotic prophylaxis remain unclear. In the current study, among 103 children treated on the AML02 protocol between October 2002 and October 2008 at St. Jude Children's Research Hospital, the authors retrospectively assessed the effect of antibiotic prophylaxis on the frequency of febrile neutropenia, clinically or microbiologically confirmed infections (including bacteremia), and antibiotic resistance, as well as on the results of nasal and rectal surveillance cultures. Initially, patients received no prophylaxis or oral cephalosporin (group A). The protocol was then amended to administer intravenous cefepime alone or intravenous vancomycin plus either oral cephalosporin, oral ciprofloxacin, or intravenous cefepime (group B). There were 334 infectious episodes. Patients in group A had a significantly greater frequency of documented infections and bacteremia (both P < .0001) (including gram-positive and gram-negative bacteremia; P = .0003 and .001, respectively) compared with patients in group B, especially viridans streptococcal bacteremia (P = .001). The incidence of febrile neutropenia without documented infection was not found to be different between the 2 groups. Five cases of bacteremia with vancomycin-resistant enterococci (VRE) occurred in group B (vs none in group A), without related mortality. Two of these cases were preceded by positive VRE rectal surveillance cultures. Outpatient intravenous antibiotic prophylaxis is feasible in children with acute myeloid leukemia and reduces the frequency of documented infection but not of febrile neutropenia. Despite the emergence of VRE bacteremia, the benefits favor antibiotic prophylaxis. Creative approaches to shorten the duration of prophylaxis and thereby minimize resistance should be explored.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, ≥85% (Vancomycin B)
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, BioReagent, suitable for plant cell culture
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vancomycin -hydrochlorid aus Streptomyces orientalis, meets USP testing specifications
USP
Ciprofloxacinhydrochlorid, United States Pharmacopeia (USP) Reference Standard
Millipore
Vancomycin-Supplement, suitable for microbiology
Vancomycin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin-Hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacinhydrochlorid für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard