Merck
  • Preventing intraperitoneal adhesions with linezolid and hyaluronic acid/carboxymethylcellulose: a comparative study in cecal abrasion model.
Preventing intraperitoneal adhesions with linezolid and hyaluronic acid/carboxymethylcellulose: a comparative study in cecal abrasion model.
American journal of surgery (2014-05-13)
Kasim Caglayan, Bulent Gungor, Hamza Cinar, Nilsen Y Erdogan, Bulent Koca
ZUSAMMENFASSUNG
We aimed to compare the effectiveness of linezolid in preventing intraperitoneal adhesions with hyaluronic acid + carboxymethylcellulose (Seprafilm). Thirty rats were divided randomly into 3 groups: Group I (control), untreated; Group II (Seprafilm); and Group III (linezolid). All rats were sacrificed on the 14th day after surgery. Macroscopic adhesion, inflammation, and fibrosis were evaluated. The multiple comparisons between groups showed a statistically significant difference for adhesion. There were statistically significant differences between Group I and II and I and III, but no statistically significant difference between Group II and III. The multiple comparisons between the groups showed a statistically significant difference for inflammation and fibrosis. For inflammation and fibrosis, there was a statistically significant difference between Group I and II and I and III, but no statistically significant difference between Group II and III. The efficiency of linezolid in reducing the formation of intraperitoneal adhesions was statistically significant compared with the control group.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, low viscosity
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, High viscosity
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, viscosity 2500-6000 cP 
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, medium viscosity
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Rinderaugen-Glaskörper
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 8,000-15,000
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, ultra high viscosity, highly purified
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 750,000-1,000,000
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Xylazin, ≥99%
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 130,000-150,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 150,000-300,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 1,200
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 500,000-750,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 50,000-90,000