Merck
Carbon and chlorine isotope fractionation during Fenton-like degradation of trichloroethene.
Chemosphere (2014-05-31)
Yunde Liu, Yiqun Gan, Aiguo Zhou, Cunfu Liu, Xiaoqian Li, Tingting Yu
ZUSAMMENFASSUNG
Dual isotope approach has been proposed as a viable tool for characterizing and assessing in situ contaminant transformation, however, little data is currently available on its applicability to chlorinated ethenes. This study determined carbon and chlorine isotope fractionation during Fenton-like degradation of trichloroethene (TCE). Carbon and chlorine isotope enrichment factors were εC=-2.9 ± 0.3‰ and εCl=-0.9 ± 0.1‰, respectively. An observed small secondary chlorine isotope effect (AKIECl=1.001) was consistent with an initial transformation by adding hydroxyl radicals (OH) to CC bonds without cleavage of CCl bonds. The relative change in carbon and chlorine isotope ratios (Δ=Δδ(13)C/Δδ(37)Cl) was calculated to be 3.1 ± 0.2, approximately equal to the ratio of chlorine and carbon isotope enrichment factors (εC/εCl=3.2). The similarity of the Δ (or εC/εCl) values between Fenton-like degradation and microbial reductive dechlorination of TCE was observed, indicating that application of solely dual isotope approach may be limited in distinguishing the two transformation pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Trichlorethylen, ACS reagent, ≥99.5%
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Chlor, ≥99.5%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Supelco
Trichlorethylen, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichlorethylen, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.
Supelco
Trichlorethylen -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Chlor, puriss., ≥99.8%
USP
Lösungsmittelrückstände Klasse 2 - Trichlorethylen, United States Pharmacopeia (USP) Reference Standard