Merck
  • Conserved glycine 232 in the ligand channel of ba3 cytochrome oxidase from Thermus thermophilus.
Conserved glycine 232 in the ligand channel of ba3 cytochrome oxidase from Thermus thermophilus.
Biochemistry (2014-06-18)
William McDonald, Chie Funatogawa, Yang Li, Ying Chen, Istvan Szundi, James A Fee, C David Stout, Olöf Einarsdóttir
ZUSAMMENFASSUNG
Knowing how the protein environment modulates ligand pathways and redox centers in the respiratory heme-copper oxidases is fundamental for understanding the relationship between the structure and function of these enzymes. In this study, we investigated the reactions of O2 and NO with the fully reduced G232V mutant of ba3 cytochrome c oxidase from Thermus thermophilus (Tt ba3) in which a conserved glycine residue in the O2 channel of the enzyme was replaced with a bulkier valine residue. Previous studies of the homologous mutant of Rhodobacter sphaeroides aa3 cytochrome c oxidase suggested that the valine completely blocked the access of O2 to the active site [Salomonsson, L., et al. (2004) Proc. Natl. Acad. Sci. U.S.A. 101, 11617-11621]. Using photolabile O2 and NO carriers, we find by using time-resolved optical absorption spectroscopy that the rates of O2 and NO binding are not significantly affected in the Tt ba3 G232V mutant. Classical molecular dynamics simulations of diffusion of O2 to the active site in the wild-type enzyme and G232V mutant show that the insertion of the larger valine residue in place of the glycine appears to open up other O2 and NO exit/entrance pathways that allow these ligands unhindered access to the active site, thus compensating for the larger valine residue.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
L-Valin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Glycin -hydrochlorid, ≥99% (HPLC)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
SAFC
Glycin
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
L-Valin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Valin
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
L-Valin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycin, 99%, FCC
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin 1 M -Lösung
Supelco
L-Valin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method