Merck
  • Impact of P-glycoprotein on blood-retinal barrier permeability: comparison of blood-aqueous humor and blood-brain barrier using mdr1a knockout rats.
Impact of P-glycoprotein on blood-retinal barrier permeability: comparison of blood-aqueous humor and blood-brain barrier using mdr1a knockout rats.
Investigative ophthalmology & visual science (2014-07-06)
Shinobu Fujii, Chikako Setoguchi, Kouichi Kawazu, Ken-ichi Hosoya
ZUSAMMENFASSUNG
The purpose of this study was to clarify the impact of P-glycoprotein (P-gp) on blood-retinal barrier (BRB) and blood-aqueous humor barrier (BAB) permeability, in contrast to blood-brain barrier (BBB) permeability. Permeabilities of six compounds, including P-gp substrates (quinidine, digoxin, and verapamil), were investigated in wild-type and mdr1a knockout rats using retinal, aqueous humor, and brain uptake index (RUI, AHUI, and BUI, respectively) methods and integration plot analysis. In both rat strains, quinidine, digoxin, and verapamil were transported by P-gp across each barrier; however, the impact of P-gp on retinal uptake of quinidine and verapamil was less pronounced than that on brain uptake. The apparent influx permeability clearance (Kin) values of verapamil in retina obtained from wild-type and knockout rats were similar (0.824 ± 0.201 and 0.849 ± 0.980 mL/min·g retina, respectively; mean ± SD; n = 3 rats). The Kin in aqueous humor and brain obtained from knockout rats was, respectively, 3-fold and 12-fold higher than that of wild-type (P < 0.05). In P-gp-deficient conditions, the RUI and AHUI of quinidine, digoxin, and verapamil, as well as the BUI of quinidine and digoxin, were decreased by P-gp inhibitors. However, the BUI of verapamil was not changed by P-gp inhibitors. Results suggest that carrier-mediated influx transporters exist in the blood-ocular barriers and that the function of verapamil influx transporters is markedly different between the retina and brain. In both rat strains, P-gp operates in the blood-ocular barriers, and the impact of P-gp on BRB permeability to quinidine and verapamil is lower than that on BBB permeability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil -hydrochlorid, ≥99% (titration), powder
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Chinidin, anhydrous
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
USP
Verapamil -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Chinidin, crystallized, ≥98.0% (dried material, NT)
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-Verapamil -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material