Merck
  • Microfunnel-supported liquid-phase microextraction: application to extraction and determination of Irgarol 1051 and diuron in the Persian Gulf seawater samples.
Microfunnel-supported liquid-phase microextraction: application to extraction and determination of Irgarol 1051 and diuron in the Persian Gulf seawater samples.
Journal of chromatography. A (2014-07-14)
Abolfazl Saleh, Neda Sheijooni Fumani, Saeideh Molaei
ZUSAMMENFASSUNG
In the present work, microfunnel-supported liquid-phase microextraction method (MF-LPME) based on applying low density organic solvent was developed for the determination of antifoulings (Irgarol 1051, diuron and 3,4-dichloroaniline) from seawater samples. In this method, home-designed MF device was used for facile loading and retrieving of organic solvent during the extraction procedure. The extraction was carried out with introduction of 400 μL of toluene via syringe into the MF device placed on the surface of sample solution (300 mL) containing analytes. After the extraction, extractant layer was narrowed into the capillary part of MF by pushing the device inside the sample and withdrawn by using a syringe to evaporate by nitrogen purging. The residual redissolved into 50 μL methanol, diluted to 100 μL with deionized water and injected into the high performance liquid chromatography with UV detection (HPLC-UV). Several factors influencing the extraction such as the type and volume of extraction solvent, sample pH, extraction time and ionic strength were investigated and optimized. Under the optimized conditions, the limits of detection in seawater were 1.4, 4.8 and 1.0 ng L(-1) for 3,4-dichloroaniline (DCA), diuron and Irgarol 1051, respectively. Enrichment factors were obtained 333, 150 and 373 for DCA, diuron and Irgarol 1051, respectively. The precision of the technique was evaluated in terms of repeatability which was less than 12.0% (n=5). The applicability of the proposed method was evaluated by the extraction and determination of antifoulings from seawater samples collected from harbors of Bushehr located in northern Persian Gulf coast.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,4-Dichloranilin, 98%
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Supelco
Toluol -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Irgarol®, PESTANAL®, analytical standard
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Diuron, PESTANAL®, analytical standard
Supelco
Toluol-Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL