Merck
Innate immune cell CD45 regulates lymphopenia-induced T cell proliferation.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-13)
Amy E Saunders, Yaein A Shim, Pauline Johnson
ZUSAMMENFASSUNG
The leukocyte-specific tyrosine phosphatase, CD45, severely impacts T cell development and activation by modulating TCR signaling. CD45-deficient (CD45KO) mice have reduced peripheral T cell numbers where CD8 T cells are underrepresented. In this article, we show that CD45KO mice are unable to support efficient homeostatic proliferation, affecting CD8 T cells more than CD4 T cells. Using CD45-RAG1 double-deficient (45RAGKO) mice, we show that lymphopenia-induced proliferation (LIP) of CD45-sufficient T cells is defective in a host environment lacking CD45 on innate immune cells. We identify two deficiencies in the 45RAGKO mice that affect LIP. One involves CD11c(+) cells and the second the production of IL-7 by lymphoid stromal cells. CD45KO dendritic cells were not defective in foreign Ag-induced T cell proliferation, yet CD45KO CD11c(+) cells were unable to rescue the spontaneous LIP in the 45RAGKO mice. This was in contrast with the CD45-sufficient CD11c(+) cells that partially rescued this spontaneous proliferation and did so without affecting IL-7 levels. The absence of CD45 also led to reduced IL-7 production by lymphoid stromal cells, suggesting an indirect effect of CD45 on innate immune cells in influencing IL-7 production by lymphoid stromal cells. These findings demonstrate a novel role for CD45 on innate immune cells in promoting lymphopenia-induced T cell proliferation and suggest that innate immune cells may communicate with stromal cells to regulate IL-7 production.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
SAFC
L-Glutamin
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland