Merck
Hepatocellular carcinoma in thalassaemia: an update of the Italian Registry.
British journal of haematology (2014-07-06)
Caterina Borgna-Pignatti, Maria Chiara Garani, Gian Luca Forni, Maria Domenica Cappellini, Elena Cassinerio, Carmelo Fidone, Vincenzo Spadola, Aurelio Maggio, Gaetano Restivo Pantalone, Antonio Piga, Filomena Longo, Maria Rita Gamberini, Paolo Ricchi, Silvia Costantini, Domenico D'Ascola, Paolo Cianciulli, Maria Eliana Lai, Maria Paola Carta, Angela Ciancio, Paola Cavalli, Maria Caterina Putti, Susanna Barella, Giovanni Amendola, Saveria Campisi, Marcello Capra, Vincenzo Caruso, Grazia Colletta, Stefano Volpato
ZUSAMMENFASSUNG
The risk of developing hepatocellular carcinoma (HCC) in patients with thalassaemia is increased by transfusion-transmitted infections and haemosiderosis. All Italian Thalassaemia Centres use an ad hoc form to report all diagnoses of HCC to the Italian Registry. Since our last report, in 2002, up to December 2012, 62 new cases were identified, 52% of whom were affected by thalassaemia major (TM) and 45% by thalassaemia intermedia (TI). Two had sickle-thalassaemia (ST). The incidence of the tumour is increasing, possibly because of the longer survival of patients and consequent longer exposure to the noxious effects of the hepatotropic viruses and iron. Three patients were hepatitis B surface antigen-positive, 36 patients showed evidence of past infection with hepatitis B virus (HBV). Fifty-four patients had antibodies against hepatitis C virus (HCV), 43 of whom were HCV RNA positive. Only 4 had no evidence of exposure either to HCV or HBV. The mean liver iron concentration was 8 mg/g dry weight. Therapy included chemoembolization, thermoablation with radiofrequency and surgical excision. Three patients underwent liver transplant, 21 received palliative therapy. As of December 2012, 41 patients had died. The average survival time from HCC detection to death was 11·5 months (1·4-107·2 months). Ultrasonography is recommended every 6 months to enable early diagnosis of HCC, which is crucial to decrease mortality.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eisen, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Ferritin aus Pferdemilz, Type I, saline solution
Sigma-Aldrich
Eisen, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Carbonyleisen, ≥97% Fe basis
Sigma-Aldrich
Eisen, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Eisen, powder, −325 mesh, 97%
Sigma-Aldrich
Eisen, chips, 99.98% trace metals basis
Sigma-Aldrich
Eisen, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Eisen, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Eisen, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Eisen, foil, 100x100mm, thickness 0.25mm, hard, 99.5%
Eisen, foil, 100x100mm, thickness 0.1mm, hard, 99.5%
Eisen, IRMM®, certified reference material, 0.5 mm wire
Eisen, foil, 300x300mm, thickness 0.1mm, hard, 99.5%
Eisen, rod, 100mm, diameter 2.0mm, as drawn, 99.95%
Eisen, rod, 50mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%
Eisen, rod, 200mm, diameter 25mm, as drawn, 98+%
Eisen, rod, 100mm, diameter 100mm, as drawn, armcO« soft ingot 99.8%
Eisen, foil, 150x150mm, thickness 1.5mm, as rolled, 99.5%
Eisen, rod, 200mm, diameter 9.5mm, as drawn, soft ingot 99.8+%
Eisen, rod, 200mm, diameter 3.0mm, as drawn, 99.99+%
Eisen, rod, 200mm, diameter 6.35mm, as drawn, soft ingot 99.8%
Eisen, foil, 100x100mm, thickness 1.0mm, as rolled, 99.5%
Eisen, tube, 200mm, outside diameter 3.2mm, inside diameter 2.2mm, wall thickness 0.5mm, as drawn, 99.8%
Eisen, foil, 25x25mm, thickness 0.9mm, as rolled, 99.5%
Eisen, rod, 50mm, diameter 5.0mm, as drawn, 99.99+%
Eisen, rod, 500mm, diameter 9.5mm, as drawn, soft ingot 99.8+%
Eisen, foil, 25x25mm, thickness 0.5mm, hard, 99.5%