Merck
Evidence for fatty acids mediating CL 316,243-induced reductions in blood glucose in mice.
American journal of physiology. Endocrinology and metabolism (2014-08-07)
Rebecca E K MacPherson, Laura Castellani, Marie-Soleil Beaudoin, David C Wright
ZUSAMMENFASSUNG
CL 316,243, a β3-adrenergic agonist, was developed as an antiobesity and diabetes drug and causes rapid decreases in blood glucose levels in mice. The mechanisms mediating this effect have not been fully elucidated; thus, the purpose of the current study was to examine the role of fatty acids and interleukin-6, reputed mediators of insulin secretion, in this process. To address this question, we used physiological and pharmacological approaches in combination with knockout mouse models. CL 316,243 treatment in male C57BL6 mice increased plasma fatty acids, glycerol, interleukin-6, and insulin and reduced blood glucose concentrations 2 h following injections. The ability of CL 316,243 to increase insulin and fatty acids and reduce glucose was preserved in interleukin-6-deficient mice. CL 316,243-induced drops in blood glucose occurred in parallel with increases in circulating fatty acids but prior to increases in plasma interleukin-6. CL 316,243-mediated increases in plasma insulin levels and reductions in blood glucose were attenuated when mice were pretreated with the lipase inhibitor nicotinic acid or in whole body adipose tissue triglyceride lipase knockout mice. Collectively, our findings demonstrate an important role for fatty acids in mediating the effects of CL 316,243 in mice. Not only do our results provide new insight into the mechanisms of action of CL 316,243, but they also hint at an unappreciated aspect of adipose tissue -pancreas cross-talk.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinsäure, ≥99.5% (HPLC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Nicotinsäure, ≥98%
Supelco
Nicotinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Nicotinsäure, analytical standard
Supelco
Glycerin, analytical standard
USP
Niacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triglycerid- und freie Glycerin-Kits und -Reagenzien, 1 kit sufficient for 1000 reactions
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Nicotinsäure, meets USP testing specifications
Sigma-Aldrich
Nicotinsäure Natriumsalz, 98%
Nicotinsäure, European Pharmacopoeia (EP) Reference Standard