Merck
  • Effects of acute glucocorticoid blockade on metabolic dysfunction in patients with Type 2 diabetes with and without fatty liver.
Effects of acute glucocorticoid blockade on metabolic dysfunction in patients with Type 2 diabetes with and without fatty liver.
American journal of physiology. Gastrointestinal and liver physiology (2014-08-12)
D P Macfarlane, P J Raubenheimer, T Preston, C D Gray, M E Bastin, I Marshall, J P Iredale, R Andrew, B R Walker
ZUSAMMENFASSUNG
To investigate the potential of therapies which reduce glucocorticoid action in patients with Type 2 diabetes we performed a randomized, double-blinded, placebo-controlled crossover study of acute glucocorticoid blockade, using the glucocorticoid receptor antagonist RU38486 (mifepristone) and cortisol biosynthesis inhibitor (metyrapone), in 14 men with Type 2 diabetes. Stable isotope dilution methodologies were used to measure the rates of appearance of glucose, glycerol, and free fatty acids (FFAs), including during a low-dose (10 mU·m⁻² ·min⁻¹) hyperinsulinemic clamp, and subgroup analysis was conducted in patients with high or low liver fat content measured by magnetic resonance spectroscopy (n = 7/group). Glucocorticoid blockade lowered fasting glucose and insulin levels and improved insulin sensitivity of FFA and glycerol turnover and hepatic glucose production. Among this population with Type 2 diabetes high liver fat was associated with hyperinsulinemia, higher fasting glucose levels, peripheral and hepatic insulin resistance, and impaired suppression of FFA oxidation and FFA and glycerol turnover during hyperinsulinemia. Glucocorticoid blockade had similar effects in those with and without high liver fat. Longer term treatments targeting glucocorticoid action may be useful in Type 2 diabetes with and without fatty liver.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Hydrocortison -Lösung, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Cortisol -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mifepriston, ≥98%
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Sigma-Aldrich
2-Methyl-1,2-di-3-Pyridyl-1-Propanon, 96%
Sigma-Aldrich
Metyrapon, ≥98% (HPLC), solid
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard