Merck
  • The reversible P2Y12 inhibitor ticagrelor inhibits metastasis and improves survival in mouse models of cancer.
The reversible P2Y12 inhibitor ticagrelor inhibits metastasis and improves survival in mouse models of cancer.
International journal of cancer (2014-05-07)
Simon Gebremeskel, Terry LeVatte, Robert S Liwski, Brent Johnston, Michael Bezuhly
ZUSAMMENFASSUNG
Tumor cells use activated platelets to promote their proliferation and metastatic potential. Because platelet activation is largely mediated through ADP engagement of purinergic P2Y12 receptors on platelets, we investigated the potential of the reversible P2Y12 inhibitor ticagrelor, a clinical agent used in the prevention of cardiovascular and cerebrovascular events, to inhibit tumor adhesion and metastasis. In B16-F10 melanoma intravenous and intrasplenic metastasis models, mice treated with a clinical dose of ticagrelor (10 mg/kg) exhibited marked reductions in lung (84%) and liver (86%) metastases. Furthermore, ticagrelor treatment improved survival compared to saline-treated animals. A similar effect was observed in a 4T1 breast cancer model, with reductions in lung (55%) and bone marrow (87%) metastases following ticagrelor treatment. In vitro, B16-F10 cells exhibited decreased interaction with platelets from ticagrelor-treated mice compared to saline-treated mice, an effect similar to that observed with blockade of glycoprotein IIbIIIa. Similarly, B16-F10 cells co-incubated with platelets from ticagrelor-treated mice exhibited reduced adhesion to endothelial monolayers compared to those co-incubated with platelets from saline-treated animals, an effect also observed in vivo. Interestingly, pretreatment of endothelial monolayers with ticagrelor did not result in reduced tumor cell adhesion. These findings support a role for P2Y12-mediated platelet activation in promoting metastases, and provide proof-of-concept for the clinical use of ticagrelor in the prevention of tumor metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosin, ≥99%
Sigma-Aldrich
Adenosin, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosin
Supelco
Adenosin, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosin, European Pharmacopoeia (EP) Reference Standard