Merck
  • Prospective study of polydimethylsiloxane vs dextranomer/hyaluronic acid injection for treatment of vesicoureteral reflux.
Prospective study of polydimethylsiloxane vs dextranomer/hyaluronic acid injection for treatment of vesicoureteral reflux.
The Journal of urology (2014-06-15)
Katherine Moore, Stéphane Bolduc
ZUSAMMENFASSUNG
Endoscopic injection of a bulking agent is becoming a first-line treatment for low grade vesicoureteral reflux. We prospectively compared the efficacy of 2 such products commercially available in Canada. A total of 275 patients with documented grade I to V vesicoureteral reflux were prospectively enrolled in a comparative study between April 2005 and February 2011 to be randomly treated endoscopically with either polydimethylsiloxane (Macroplastique®) or dextranomer/hyaluronic acid copolymer (Deflux®). Of the ureters 202 were treated with polydimethylsiloxane and 197 with dextranomer/hyaluronic acid copolymer. Patients were followed with voiding cystourethrography at 3 months and renal ultrasonography at 3 months and at 1 year. Median followup was 4.3 years. The primary outcome was surgical success (resolution vs nonresolution), and secondary outcomes included occurrence of adverse events. Vesicoureteral reflux was fully corrected in 182 of 202 ureters (90%) treated with polydimethylsiloxane, compared to 159 of 197 (81%) treated with dextranomer/hyaluronic acid copolymer (p <0.05). Obstruction was found in 5 ureters. Univariate and multivariate analyses did not allow identification of any characteristics that could explain the significant difference in the success rates except for the type of product used. We present the largest known prospective evaluation comparing 2 bulking agents for the treatment of vesicoureteral reflux. Endoscopic injection of polydimethylsiloxane resulted in a better success rate than dextranomer/hyaluronic acid copolymer. The rate of resolution obtained with the latter is lower than those previously published due to the inclusion of high grade reflux.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Siliconöl, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Siliconöl
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Siliconöl, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Siliconöl, viscosity 50 cSt (25 °C)
Siliconöl, for melting point and boiling point apparatuses
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Rinderaugen-Glaskörper
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 8,000-15,000
Sigma-Aldrich
Siliconöl, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, 25,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 670,000
Sigma-Aldrich
Hyaluronsäure Natriumsalz aus Streptococcus equi sp., mol wt 750,000-1,000,000
Sigma-Aldrich
Siliconöl, viscosity 10,000 cSt (25 °C)
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000