Merck
  • Metabolic profiling of synovial fluid in a unilateral ovine model of anterior cruciate ligament reconstruction of the knee suggests biomarkers for early osteoarthritis.
Metabolic profiling of synovial fluid in a unilateral ovine model of anterior cruciate ligament reconstruction of the knee suggests biomarkers for early osteoarthritis.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-10-07)
Beata Mickiewicz, Bryan J Heard, Johnny K Chau, May Chung, David A Hart, Nigel G Shrive, Cyril B Frank, Hans J Vogel
ZUSAMMENFASSUNG
Joint injuries and subsequent osteoarthritis (OA) are the leading causes of chronic joint disease. In this work, we explore the possibility of applying magnetic resonance spectroscopy-based metabolomics to detect host responses to an anterior cruciate ligament (ACL) reconstruction injury in synovial fluid in an ovine model. Using multivariate statistical analysis, we were able to distinguish post-injury joint samples (ACL and sham surgery) from the uninjured control samples, and as well the ACL surgical samples from sham surgery. In all samples there were 65 metabolites quantified, of which six could be suggested as biomarkers for early post-injury degenerative changes in the knee joints: isobutyrate, glucose, hydroxyproline, asparagine, serine, and uridine. Our results raise a cautionary note indicating that surgical interventions into the knee can result in metabolic alterations that need to be distinguished from those caused by the early onset of OA. Our findings illustrate the potential application of metabolomics as a diagnostic and prognostic tool for detection of injuries to the knee joint. The ability to detect a unique pattern of metabolic changes in the synovial fluid of sheep offers the possibility of extending the approach to precision medicine protocols in patient populations in the future.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Uridin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Uridin, ≥99%
Sigma-Aldrich
L-Asparagin, ≥98% (HPLC)
Sigma-Aldrich
Trans-4-Hydroxy-L-Prolin, ≥99%
Sigma-Aldrich
L-Asparagin, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Trans-4-Hydroxy-L-Prolin, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Uridin, BioUltra, ≥99%
Sigma-Aldrich
Trans-4-Hydroxy-L-Prolin, BioXtra, ≥99.0% (NT)
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trans-4-Hydroxy-L-Prolin, analytical standard
Supelco
L-Asparagin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Serin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland