Merck
  • Modelling of the growth/no growth interface of Wallemia sebi and Eurotium herbariorum as a function of pH, aw and ethanol concentration.
Modelling of the growth/no growth interface of Wallemia sebi and Eurotium herbariorum as a function of pH, aw and ethanol concentration.
International journal of food microbiology (2014-10-16)
N Deschuyffeleer, A Vermeulen, J Daelman, E Castelein, M Eeckhout, F Devlieghere
ZUSAMMENFASSUNG
High sugar products (sugar content > 50%) are generally considered to be stable against all forms of microbial spoilage during a prolonged shelf life of several months. However, one specific subgroup of micro-organisms, the xerophilic moulds, can develop quite fast on the surface of food products with a reduced water activity (< 0.85). The chance whether these xerophilic moulds are able to grow on the food product depends on the combination of intrinsic factors (e.g., water activity and pH) and the storage conditions (e.g., temperature). This study examines the development of growth/no growth models for the xerophilic moulds Wallemia sebi and Eurotium herbariorum in a sugar rich broth. Growth/no growth models are predictive models that are designed to give a prediction about the probability of growth of a spoilage micro-organism under a specific set of environmental conditions. In this research, a water activity between 0.75 and 0.90, a pH between 5.0 and 6.2, an ethanol concentration between 0% and 5% (g EtOH/g H2O) and their interactions were tested. The inoculated media were stored at 22 °C (± 1 °C) during a prolonged test period (up to 120 days). The obtained models were also validated in a chocolate-based food product (ganache). The resulting growth/no growth models show that the growth of W. sebi and E. herbariorum can be inhibited for a prolonged time (> 3 months) if an ethanol concentration of 5% on the water phase is present in the food product, irrespective of water activity values between 0.89 and 0.755. The necessary amount of ethanol for shorter shelf lives can be calculated with the models that were built. Although the models have not been validated thoroughly in actual food products, the preliminary results that were obtained by testing the model on a ganache indicated that the models are capable of delivering safe predictions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Wasser
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Wasser, BioPerformance Certified
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Wasser, for ultratrace analysis, ACS reagent
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol