Merck
  • The laboratory performance of the enFLOW(®) , buddy lite(™) and ThermoSens(®) fluid warmers.
The laboratory performance of the enFLOW(®) , buddy lite(™) and ThermoSens(®) fluid warmers.
Anaesthesia (2014-11-13)
H J Kim, S M Yoo, J S Jung, S H Lee, K Sun, H S Son
ZUSAMMENFASSUNG
We measured heating of isotonic saline by three fluid warmers in six experiments: saline at 5 °C or 20 °C delivered at 30, 50 or 100 ml.min(-1) . At the three flow rates, the enFLOW(®) , buddy lite(™) and ThermoSens(®) systems heated 5 °C saline to mean (SD) temperatures of: 41.1 (0.5) °C, 37.7 (0.6) °C and 39.1 (0.6) °C; to 40.3 (0.8) °C, 33.9 (1.6) °C and 39.3 (0.7) °C; and to 37.1 (0.8) °C, 24.0 (1.3) °C and 37.6 (1.0) °C, respectively, p < 0.0001 for each experiment. The mean (SD) times taken to heat 5 °C saline were: 16.6 (1.7) s, 258.4 (58.9) s and 134.2 (79.6) s; 16.9 (1.8) s, 256.2 (62.2) s and 182.5 (74.5) s; and 21.5 (1.5) s, 275.9 (49.3) s and 313.5 (18.0) s, respectively, p < 0.0003 for each experiment. The results for saline at 20 °C were similar. The enFLOW system heated saline above 36 °C faster than the ThermoSens system, whereas the buddy lite often failed to achieve 36 °C.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis