Merck
  • The effects of anesthesia, muscle paralysis, and ventilation on the lung evaluated by lung diffusion for carbon monoxide and pulmonary surfactant protein B.
The effects of anesthesia, muscle paralysis, and ventilation on the lung evaluated by lung diffusion for carbon monoxide and pulmonary surfactant protein B.
Anesthesia and analgesia (2014-11-02)
Fabiano Di Marco, Daniele Bonacina, Emanuele Vassena, Erik Arisi, Anna Apostolo, Cristina Banfi, Stefano Centanni, Piergiuseppe Agostoni, Roberto Fumagalli
ZUSAMMENFASSUNG
An increased alveolar-arterial oxygen tension difference is frequent in anesthetized patients. In this study, we evaluated the effect on the lung of anesthesia, muscle paralysis, and a brief course of mechanical ventilation. Lung diffusion for carbon monoxide (DLCO), including pulmonary capillary blood volume (Vc) and conductance of the alveolar-capillary membrane (DM), and pulmonary surfactant protein type B (a marker of alveolar damage) were measured in 45 patients without pulmonary disease undergoing extrathoracic surgery. Anesthesia, muscle paralysis, and mechanical ventilation led to impairment of gas exchange, with a reduction of DLCO values immediately after anesthetic induction due to a concomitant reduction of both DM and Vc. While changes in DM were due to the reduction of lung volume, changes in Vc were not limited to volume loss, since the Vc/alveolar volume ratio decreased significantly. Although DLCO and its components decreased immediately after induction, none of the values decreased further at 1 and 3 hours. Surfactant protein type B, however, was unchanged immediately after anesthesia but increased at 1 hour after induction and further increased after 3 hours of anesthesia. The level of alveolar damage correlated with the reduction of lung perfusion and lung dynamic strain (i.e., ratio between tidal volume and end-expiratory lung volume). A brief course of anesthesia and controlled ventilation leads to: (1) alveolar damage, which is correlated with lung strain and perfusion, and (2) impaired gas exchange mainly due to volume loss but also to reduced aerated lung perfusion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kohlenstoffmonoxid, ≥99.0%
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Kohlenstoff, mesoporös, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Kohlenstoff, mesoporös
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis