Merck
Spore immobilization and its analytical performance for monitoring of aflatoxin M1 in milk.
Canadian journal of microbiology (2014-11-13)
V K Singh, N A Singh, N Kumar, H V Raghu, Pradeep Kumar Sharma, K P Singh, Avinash Yadav
ZUSAMMENFASSUNG
Immobilization of Bacillus megaterium spores on Eppendorf tubes through physical adsorption has been used in the detection of aflatoxin M1 (AFM1) in milk within real time of 45 ± 5 min using visual observation of changes in a chromogenic substrate. The appearance of a sky-blue colour indicates the absence of AFM1 in milk, whereas no colour change indicates the presence of AFM1 in milk at a 0.5 ppb Codex maximum residue limit. The working performance of the immobilized spores was shown to persist for up to 6 months. Further, spores immobilized on 96-well black microtitre plates by physical adsorption and by entrapment on sensor disk showed a reduction in detection sensitivity to 0.25 ppb within a time period of 20 ± 5 min by measuring fluorescence using a microbiological plate reader through the addition of milk and fluorogenic substrate. A high fluorescence ratio indicated more substrate hydrolysis due to spore-germination-mediated release of marker enzymes of spores in the absence of AFM1 in milk; however, low fluorescence ratios indicated the presence of AFM1 at 0.25 ppb. Immobilized spores on 96-well microtitre plates and sensor disks have shown better reproducibility after storage at 4 °C for 6 months. Chromogenic assay showed 1.38% false-negative and 2.77% false-positive results while fluorogenic assay showed 4.16% false-positive and 2.77% false-negative results when analysed for AFM1 using 72 milk samples containing raw, pasteurized, and dried milk. Immobilization of spores makes these chromogenic and fluorogenic assays portable, selective, cost-effective for real-time detection of AFM1 in milk at the dairy farm, reception dock, and manufacturing units of the dairy industry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszeindiacetat, used as cell viability stain
Sigma-Aldrich
Aflatoxin M1, from Aspergillus flavus
Supelco
Aflatoxin M1 -Lösung, 0.5 μg/mL in acetonitrile, analytical standard
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard