Merck
  • Effect of atorvastatin therapy on fibrous cap thickness in coronary atherosclerotic plaque as assessed by optical coherence tomography: the EASY-FIT study.
Effect of atorvastatin therapy on fibrous cap thickness in coronary atherosclerotic plaque as assessed by optical coherence tomography: the EASY-FIT study.
Journal of the American College of Cardiology (2014-12-03)
Kenichi Komukai, Takashi Kubo, Hironori Kitabata, Yoshiki Matsuo, Yuichi Ozaki, Shigeho Takarada, Yasushi Okumoto, Yasutsugu Shiono, Makoto Orii, Kunihiro Shimamura, Satoshi Ueno, Takashi Yamano, Takashi Tanimoto, Yasushi Ino, Tomoyuki Yamaguchi, Hirata Kumiko, Atsushi Tanaka, Toshio Imanishi, Hideharu Akagi, Takashi Akasaka
ZUSAMMENFASSUNG
The detailed mechanism of plaque stabilization by statin therapy is not fully understood. The aim of this study was to assess the effect of lipid-lowering therapy with 20 mg/day of atorvastatin versus 5 mg/day of atorvastatin on fibrous cap thickness in coronary atherosclerotic plaques by using optical coherence tomography (OCT). Seventy patients with unstable angina pectoris and untreated dyslipidemia were randomized to either 20 mg/day or 5 mg/day of atorvastatin therapy. OCT was performed to assess intermediate nonculprit lesions at baseline and 12-month follow-up. Serum low-density lipoprotein cholesterol level was significantly lower during therapy with 20 mg/day compared with 5 mg/day of atorvastatin (69 mg/dl vs. 78 mg/dl; p = 0.039). The increase in fibrous cap thickness was significantly greater with 20 mg/day compared with 5 mg/day of atorvastatin (69% vs. 17%; p < 0.001). The increase in fibrous cap thickness correlated with the decrease in serum levels of low-density lipoprotein cholesterol (R = -0.450; p < 0.001), malondialdehyde-modified low-density lipoprotein (R = -0.283; p = 0.029), high-sensitivity C-reactive protein (R = -0.276; p = 0.033), and matrix metalloproteinase-9 (R = -0.502; p < 0.001), and the decrease in grade of OCT-derived macrophages (R = -0.415; p = 0.003). Atorvastatin therapy at 20 mg/day provided a greater increase in fibrous cap thickness in coronary plaques compared with 5 mg/day of atorvastatin. The increase of fibrous cap was associated with the decrease in serum atherogenic lipoproteins and inflammatory biomarkers during atorvastatin therapy. (Effect of Atorvastatin Therapy on Fibrous Cap Thickness in Coronary Atherosclerotic Plaque as Assessed by Optical Coherence Tomography: The EASY-FIT Study; NCT00700037).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
C Reactive Protein from human fluids, buffered aqueous solution
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.