Merck
Metabolic inflexibility: when mitochondrial indecision leads to metabolic gridlock.
Cell (2014-12-07)
Deborah M Muoio
ZUSAMMENFASSUNG
Normal energy metabolism is characterized by periodic shifts in glucose and fat oxidation, as the mitochondrial machinery responsible for carbon combustion switches freely between alternative fuels according to physiological and nutritional circumstances. These transitions in fuel choice are orchestrated by an intricate network of metabolic and cell signaling events that enable exquisite crosstalk and cooperation between competing substrates to maintain energy and glucose homeostasis. By contrast, obesity-related cardiometabolic diseases are increasingly recognized as disorders of metabolic inflexibility, in which nutrient overload and heightened substrate competition result in mitochondrial indecision, impaired fuel switching, and energy dysregulation. This Perspective offers a speculative view on the molecular origins and pathophysiological consequences of metabolic inflexibility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glukose -Lösung, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose -Lösung, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Supelco
D-(+)-Glukose, analytical standard
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glukose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glukose, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glukose -Lösung, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard