Merck
Functionalized olefin cross-coupling to construct carbon-carbon bonds.
Nature (2014-12-19)
Julian C Lo, Jinghan Gui, Yuki Yabe, Chung-Mao Pan, Phil S Baran
ZUSAMMENFASSUNG
Carbon-carbon (C-C) bonds form the backbone of many important molecules, including polymers, dyes and pharmaceutical agents. The development of new methods to create these essential connections in a rapid and practical fashion has been the focus of numerous organic chemists. This endeavour relies heavily on the ability to form C-C bonds in the presence of sensitive functional groups and congested structural environments. Here we report a chemical transformation that allows the facile construction of highly substituted and uniquely functionalized C-C bonds. Using a simple iron catalyst, an inexpensive silane and a benign solvent under ambient atmosphere, heteroatom-substituted olefins are easily reacted with electron-deficient olefins to create molecular architectures that were previously difficult or impossible to access. More than 60 examples are presented with a wide array of substrates, demonstrating the chemoselectivity and mildness of this simple reaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Aktivkohle, powder
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Supelco
Aktivkohle, puriss. p.a., powder
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Millipore
Aktivkohle, suitable for GC
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Kohlenstoff, foil, 10x10mm, thickness 2.0mm, hOpg
Kohlenstoff, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Kohlenstoff, foil, 50x50mm, thickness 0.2mm, pyrolytic graphite, 99.99%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Kohlenstoff, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Kohlenstoff, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon