Merck
Biomaterials. Electronic dura mater for long-term multimodal neural interfaces.
Science (New York, N.Y.) (2015-01-13)
Ivan R Minev, Pavel Musienko, Arthur Hirsch, Quentin Barraud, Nikolaus Wenger, Eduardo Martin Moraud, Jérôme Gandar, Marco Capogrosso, Tomislav Milekovic, Léonie Asboth, Rafael Fajardo Torres, Nicolas Vachicouras, Qihan Liu, Natalia Pavlova, Simone Duis, Alexandre Larmagnac, Janos Vörös, Silvestro Micera, Zhigang Suo, Grégoire Courtine, Stéphanie P Lacour
ZUSAMMENFASSUNG
The mechanical mismatch between soft neural tissues and stiff neural implants hinders the long-term performance of implantable neuroprostheses. Here, we designed and fabricated soft neural implants with the shape and elasticity of dura mater, the protective membrane of the brain and spinal cord. The electronic dura mater, which we call e-dura, embeds interconnects, electrodes, and chemotrodes that sustain millions of mechanical stretch cycles, electrical stimulation pulses, and chemical injections. These integrated modalities enable multiple neuroprosthetic applications. The soft implants extracted cortical states in freely behaving animals for brain-machine interface and delivered electrochemical spinal neuromodulation that restored locomotion after paralyzing spinal cord injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Silizium, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silizium, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Silikon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Silikon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Silizium, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Silikon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Silikon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Silizium, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Silikon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Silikon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Silikon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Silikon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Silikon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Silikon, rod, 100mm, diameter 25mm, crystalline, 100%
Silikon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Silikon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Silikon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Silikon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%